Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203285.1 | NP_001190214.1 | 7 - 8 | 1097 | 132 | |
NM_180426.3 | NP_850757.1 | 7 - 8 | 916 | 132 | ||
GenBank mRNA | AB493733.1 | - | 6 - 7 | 656 | 132 | |
AJ294532.1 | - | 6 - 7 | 656 | 132 | ||
AK228148.1 | - | 7 - 8 | 897 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |