Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180433.3 | NP_850764.1 | 5 - 6 | 909 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |