Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120582.5 | NP_196119.1 | 4 - 5 | 504 | 121 | |
NM_180437.2 | NP_850768.1 | 4 - 5 | 386 | 121 | ||
NM_203003.3 | NP_974732.1 | 3 - 4 | 844 | 121 | ||
GenBank mRNA | AK229425.1 | - | 4 - 5 | 447 | 121 | |
BT044057.1 | - | 3 - 4 | 335 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |