Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120643.2 | NP_196180.1 | 1 - 2 | 253 | 65 | |
NM_180444.3 | NP_850775.1 | 1 - 2 | 253 | 65 | ||
GenBank mRNA | AK176462.1 | - | 1 - 2 | 223 | 65 | |
AK176806.1 | - | 1 - 2 | 236 | 65 | ||
AY084560.1 | - | 1 - 2 | 250 | 65 | ||
BT024471.1 | - | 1 - 2 | 88 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |