Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125707.2 | NP_001119179.1 | 10 - 11 | 1433 | 78 | |
NM_120743.3 | NP_568171.1 | 10 - 11 | 1606 | 78 | ||
NM_180452.1 | NP_850783.1 | 10 - 11 | 1432 | 78 | ||
GenBank mRNA | AF360198.1 | - | 10 - 11 | 1432 | 78 | |
AY142616.1 | - | 10 - 11 | 1321 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |