Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120743.3 | NP_568171.1 | 31 - 32 | 3589 | 131 | |
NM_180452.1 | NP_850783.1 | 31 - 32 | 3415 | 131 | ||
GenBank mRNA | AF360198.1 | - | 31 - 32 | 3415 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |