Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121446.5 | NP_196946.1 | 1 - 2 | 499 | 75 | |
NM_180487.4 | NP_850818.1 | 2 - 3 | 687 | 75 | ||
NM_203050.3 | NP_974779.1 | 2 - 3 | 477 | 75 | ||
NM_203051.3 | NP_974780.1 | 2 - 3 | 526 | 75 | ||
GenBank mRNA | AY099597.1 | - | 1 - 2 | 458 | 75 | |
BT000249.1 | - | 1 - 2 | 323 | 75 | ||
DQ086862.1 | - | 1 - 2 | 323 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |