Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121620.2 | NP_568328.1 | 2 - 3 | 357 | 78 | |
NM_180497.2 | NP_850828.1 | 2 - 3 | 442 | 78 | ||
NM_203058.3 | NP_974787.1 | 1 - 2 | 529 | 78 | ||
GenBank mRNA | AY058152.1 | - | 2 - 3 | 357 | 78 | |
AY091052.1 | - | 2 - 3 | 341 | 78 | ||
AY117359.1 | - | 1 - 2 | 255 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |