Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121620.2 | NP_568328.1 | 9 - 10 | 1136 | 73 | |
NM_180497.2 | NP_850828.1 | 9 - 10 | 1221 | 73 | ||
NM_203058.3 | NP_974787.1 | 8 - 9 | 1308 | 73 | ||
GenBank mRNA | AY058152.1 | - | 9 - 10 | 1136 | 73 | |
AY091052.1 | - | 9 - 10 | 1120 | 73 | ||
AY117359.1 | - | 8 - 9 | 1034 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |