Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121634.4 | NP_197133.1 | 7 - 8 | 1026 | 90 | |
NM_180498.3 | NP_850829.1 | 7 - 8 | 1013 | 90 | ||
GenBank mRNA | AK226906.1 | - | 7 - 8 | 952 | 90 | |
BT021989.1 | - | 7 - 8 | 915 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |