Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203404.1 | NP_001190333.1 | 5 - 6 | 1255 | 125 | |
NM_121888.3 | NP_197384.1 | 5 - 6 | 1350 | 125 | ||
NM_180519.1 | NP_850850.1 | 5 - 6 | 1223 | 125 | ||
GenBank mRNA | AF367355.1 | - | 5 - 6 | 1191 | 125 | |
AJ011612.1 | - | 5 - 6 | 1149 | 125 | ||
AY063815.1 | - | 5 - 6 | 1223 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |