Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036865.1 | NP_001031942.1 | 4 - 5 | 479 | 82 | |
NM_001343973.1 | NP_001332394.1 | 4 - 5 | 696 | 82 | ||
NM_180536.3 | NP_850867.1 | 4 - 5 | 520 | 82 | ||
NM_180537.3 | NP_850868.1 | 4 - 5 | 533 | 82 | ||
GenBank mRNA | AF000147.1 | - | 4 - 5 | 401 | 82 | |
BT029445.1 | - | 3 - 4 | 341 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |