Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331357.1 | NP_001318908.1 | 3 - 4 | 299 | 57 | |
NM_001331358.1 | NP_001321307.1 | 3 - 4 | 301 | 57 | ||
NM_001331359.1 | NP_001321308.1 | 3 - 4 | 239 | 57 | ||
NM_180602.2 | NP_850933.1 | 3 - 4 | 345 | 57 | ||
NM_180603.1 | NP_850934.1 | 3 - 4 | 204 | 57 | ||
GenBank mRNA | AK118379.1 | - | 3 - 4 | 242 | 57 | |
AY085920.1 | - | 3 - 4 | 243 | 57 | ||
AY128338.1 | - | 3 - 4 | 198 | 57 | ||
BT006316.1 | - | 2 - 3 | 150 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |