Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035925.2 | NP_001031002.1 | 12 - 13 | 1967 | 80 | |
NM_001198013.1 | NP_001184942.1 | 11 - 12 | 2105 | 80 | ||
NM_001331785.1 | NP_001320735.1 | 11 - 12 | 2301 | 80 | ||
NM_100741.4 | NP_172344.2 | 11 - 12 | 2090 | 80 | ||
NM_180610.4 | NP_850941.1 | 11 - 12 | 2094 | 80 | ||
GenBank mRNA | AF184145.1 | - | 11 - 12 | 2016 | 80 | |
AK176415.1 | - | 12 - 13 | 1914 | 80 | ||
AY081303.1 | - | 11 - 12 | 2074 | 80 | ||
BT008398.1 | - | 11 - 12 | 1860 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |