Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102082.4 | NP_173650.3 | 1 - 2 | 1282 | 112 | |
NM_180618.2 | NP_850949.1 | 2 - 3 | 1211 | 112 | ||
GenBank mRNA | AK118852.1 | - | 1 - 2 | 1200 | 112 | |
AY080662.1 | - | 2 - 3 | 1085 | 112 | ||
AY133726.1 | - | 2 - 3 | 805 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |