Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333187.1 | NP_001320614.1 | 6 - 7 | 938 | 147 | |
NM_001333188.1 | NP_001320615.1 | 6 - 7 | 938 | 147 | ||
NM_001333189.1 | NP_001320613.1 | 8 - 9 | 960 | 147 | ||
NM_001333190.1 | NP_001320616.1 | 7 - 8 | 922 | 147 | ||
NM_001333191.1 | NP_001320617.1 | 6 - 7 | 934 | 147 | ||
NM_103366.3 | NP_174903.3 | 9 - 10 | 829 | 147 | ||
NM_180625.1 | NP_850956.1 | 6 - 7 | 860 | 147 | ||
NM_180626.2 | NP_850957.1 | 6 - 7 | 1007 | 147 | ||
NM_180627.2 | NP_850958.1 | 6 - 7 | 938 | 147 | ||
GenBank mRNA | AF414941.1 | - | 6 - 7 | 850 | 147 | |
AF414942.1 | - | 6 - 7 | 860 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |