Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333628.1 | NP_001321531.1 | 2 | 1295 | 103 | |
NM_104281.3 | NP_175806.3 | 2 | 1201 | 103 | ||
NM_180632.2 | NP_850963.1 | 1 | 1481 | 103 | ||
GenBank mRNA | AY074550.1 | - | 1 | 1143 | 103 | |
X71915.1 | - | 1165 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |