Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120500.3 | NP_196038.1 | 4 - 5 | 711 | 82 | |
GenBank mRNA | AK221569.1 | - | 4 - 5 | 711 | 82 | |
AY088872.1 | - | 4 - 5 | 707 | 82 | ||
DQ446916.1 | - | 4 - 5 | 664 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |