Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342755.1 | NP_001331162.1 | 7 - 8 | 1223 | 90 | |
NM_120504.2 | NP_568135.1 | 7 - 8 | 1107 | 90 | ||
NM_203000.2 | NP_974729.1 | 7 - 8 | 1081 | 90 | ||
GenBank mRNA | AB102952.1 | - | 7 - 8 | 757 | 90 | |
FM213367.1 | - | 7 - 8 | 913 | 90 | ||
FM213368.1 | - | 7 - 8 | 816 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |