Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342755.1 | NP_001331162.1 | 9 - 10 | 1828 | 109 | |
NM_120504.2 | NP_568135.1 | 9 - 10 | 1712 | 109 | ||
NM_203000.2 | NP_974729.1 | 9 - 10 | 1686 | 109 | ||
GenBank mRNA | AB102952.1 | - | 9 - 10 | 1362 | 109 | |
FM213367.1 | - | 9 - 10 | 1518 | 109 | ||
FM213368.1 | - | 9 - 10 | 1421 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |