Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342760.1 | NP_001332032.1 | 3 - 4 | 718 | 78 | |
NM_120511.1 | NP_196049.1 | 3 - 4 | 618 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |