Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342778.1 | NP_001330248.1 | 3 - 4 | 709 | 76 | |
NM_120531.3 | NP_196069.1 | 3 - 4 | 784 | 76 | ||
GenBank mRNA | BT021123.1 | - | 3 - 4 | 512 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |