Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085058.2 | NP_001078527.1 | 12 - 13 | 5255 | 74 | |
NM_001342781.1 | NP_001331593.1 | 12 - 13 | 5260 | 74 | ||
NM_120538.2 | NP_196076.2 | 10 - 11 | 5670 | 74 | ||
GenBank mRNA | DQ335243.1 | - | 12 - 13 | 5167 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |