Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085058.2 | NP_001078527.1 | 7 - 8 | 4886 | 80 | |
NM_001342781.1 | NP_001331593.1 | 7 - 8 | 4891 | 80 | ||
NM_120538.2 | NP_196076.2 | 5 - 6 | 5301 | 80 | ||
GenBank mRNA | DQ335243.1 | - | 7 - 8 | 4798 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |