Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120599.4 | NP_196136.1 | 3 - 4 | 574 | 78 | |
GenBank mRNA | BT002335.1 | - | 2 - 3 | 216 | 78 | |
KJ138704.1 | - | 2 - 3 | 216 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |