Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120621.2 | NP_196158.1 | 3 - 4 | 563 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |