Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120656.3 | NP_196193.1 | 1 - 2 | 231 | 77 | |
NM_203007.3 | NP_974736.1 | 1 - 2 | 367 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |