Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120677.2 | NP_196214.1 | 1 | 558 | 89 | |
GenBank mRNA | AK228786.1 | - | 1 | 509 | 89 | |
AY086454.1 | - | 1 | 529 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |