Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC1BAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342857.1 | NP_001332193.1 | 3 - 4 | 1352 | 69 | |
NM_120697.3 | NP_196233.1 | 4 - 5 | 1307 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |