Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120698.5 | NP_196234.3 | 5 - 6 | 769 | 79 | |
GenBank mRNA | AY039944.1 | - | 5 - 6 | 710 | 79 | |
AY113862.1 | - | 5 - 6 | 634 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |