Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120698.5 | NP_196234.3 | 9 - 10 | 1353 | 99 | |
GenBank mRNA | AY039944.1 | - | 9 - 10 | 1294 | 99 | |
AY113862.1 | - | 9 - 10 | 1218 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |