Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120717.5 | NP_196252.1 | 4 - 5 | 589 | 68 | |
GenBank mRNA | AF372941.1 | - | 4 - 5 | 522 | 68 | |
AY113163.1 | - | 4 - 5 | 461 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |