Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120770.4 | NP_196305.1 | 1 - 2 | 566 | 107 | |
GenBank mRNA | AY035121.1 | - | 1 - 2 | 566 | 107 | |
AY113916.1 | - | 1 - 2 | 543 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |