Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATJ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120773.4 | NP_196308.1 | 7 - 8 | 849 | 75 | |
GenBank mRNA | AK228246.1 | - | 7 - 8 | 790 | 75 | |
BT006202.1 | - | 7 - 8 | 651 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |