Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342930.1 | NP_001331523.1 | 3 - 4 | 248 | 91 | |
NM_120800.4 | NP_196335.2 | 5 - 6 | 803 | 91 | ||
GenBank mRNA | AY244746.1 | - | 5 - 6 | 633 | 91 | |
FJ708772.1 | - | 5 - 6 | 448 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |