Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120811.5 | NP_196346.1 | 4 - 5 | 1422 | 89 | |
GenBank mRNA | AK226960.1 | - | 4 - 5 | 1166 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |