Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120812.4 | NP_568180.1 | 14 - 15 | 1818 | 112 | |
GenBank mRNA | AF389301.1 | - | 14 - 15 | 1582 | 112 | |
BT010130.1 | - | 14 - 15 | 1547 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |