Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120821.2 | NP_196356.1 | 4 - 5 | 1048 | 107 | |
GenBank mRNA | BT003857.1 | - | 4 - 5 | 1048 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |