Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342977.1 | NP_001318505.1 | 2 | 845 | 95 | |
NM_120889.1 | NP_196424.1 | 1 | 668 | 95 | ||
GenBank mRNA | BT029995.1 | - | 1 | 668 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |