Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335740.1 | NP_001324767.1 | 7 - 8 | 605 | 68 | |
NM_127700.4 | NP_565505.1 | 7 - 8 | 536 | 68 | ||
GenBank mRNA | AF324701.2 | - | 7 - 8 | 523 | 68 | |
AF326895.1 | - | 7 - 8 | 523 | 68 | ||
AF339713.1 | - | 7 - 8 | 508 | 68 | ||
AJ516948.1 | - | 7 - 8 | 508 | 68 | ||
BT000757.1 | - | 7 - 8 | 523 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |