Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335751.1 | NP_001324104.1 | 1 - 2 | 281 | 64 | |
NM_127712.3 | NP_179735.1 | 1 - 2 | 203 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |