Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127713.4 | NP_179736.1 | 15 - 16 | 2192 | 79 | |
GenBank mRNA | AF424556.1 | - | 15 - 16 | 2041 | 79 | |
AY099690.1 | - | 15 - 16 | 2084 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |