Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SFH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036312.2 | NP_001031389.1 | 10 - 11 | 1407 | 112 | |
NM_001335761.1 | NP_001318263.1 | 10 - 11 | 1274 | 112 | ||
NM_001335763.1 | NP_001323475.1 | 10 - 11 | 1203 | 112 | ||
NM_127726.2 | NP_565514.1 | 10 - 11 | 1288 | 112 | ||
GenBank mRNA | AK176420.1 | - | 10 - 11 | 1254 | 112 | |
AY057587.1 | - | 10 - 11 | 1290 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |