Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036312.2 | NP_001031389.1 | 8 - 9 | 1300 | 62 | |
NM_001202642.2 | NP_001189571.1 | 8 - 9 | 1300 | 62 | ||
NM_001335761.1 | NP_001318263.1 | 8 - 9 | 1167 | 62 | ||
NM_001335762.1 | NP_001323476.1 | 8 - 9 | 1181 | 62 | ||
NM_001335763.1 | NP_001323475.1 | 8 - 9 | 1096 | 62 | ||
NM_127726.2 | NP_565514.1 | 8 - 9 | 1181 | 62 | ||
GenBank mRNA | AK176420.1 | - | 8 - 9 | 1147 | 62 | |
AY057587.1 | - | 8 - 9 | 1183 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |