Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127731.3 | NP_179754.1 | 3 - 4 | 693 | 80 | |
GenBank mRNA | AY057644.1 | - | 3 - 4 | 644 | 80 | |
AY113014.1 | - | 2 - 3 | 528 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |