Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124885.1 | NP_001118357.1 | 6 - 7 | 687 | 116 | |
NM_001335777.1 | NP_001323597.1 | 5 - 6 | 659 | 116 | ||
NM_001335778.1 | NP_001323598.1 | 6 - 7 | 559 | 116 | ||
NM_127757.3 | NP_565521.1 | 6 - 7 | 816 | 116 | ||
GenBank mRNA | AB071852.1 | - | 6 - 7 | 560 | 116 | |
AK229492.1 | - | 6 - 7 | 711 | 116 | ||
AY084960.1 | - | 6 - 7 | 669 | 116 | ||
BT024593.1 | - | 6 - 7 | 560 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |