Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127766.3 | NP_565524.1 | 1 - 2 | 430 | 83 | |
GenBank mRNA | AF133717.1 | - | 1 - 2 | 355 | 83 | |
AY084392.1 | - | 1 - 2 | 379 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |