Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161056.1 | NP_001154528.1 | 4 - 5 | 957 | 97 | |
NM_001335800.1 | NP_001324584.1 | 5 - 6 | 536 | 97 | ||
NM_127820.4 | NP_179840.2 | 4 - 5 | 1199 | 97 | ||
GenBank mRNA | AF211171.1 | - | 4 - 5 | 958 | 97 | |
BT033098.1 | - | 4 - 5 | 429 | 97 | ||
EU078686.1 | - | 4 - 5 | 414 | 97 | ||
EU078687.1 | - | 4 - 5 | 429 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |