Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127845.4 | NP_179865.1 | 2 - 3 | 662 | 153 | |
GenBank mRNA | AK229218.1 | - | 2 - 3 | 579 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |