Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202652.1 | NP_001189581.1 | 3 - 4 | 189 | 116 | |
NM_001335827.1 | NP_001325019.1 | 6 - 7 | 695 | 116 | ||
NM_127857.5 | NP_179876.1 | 6 - 7 | 812 | 116 | ||
NM_179702.1 | NP_850033.1 | 6 - 7 | 615 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |